Order Entry
Poland
ContactUsLinkComponent
Tetramethylsilane ACS for NMR spectroscopy
Tetramethylsilane ACS for NMR spectroscopy
Certificates
undefined
Tetramethylsilane ACS for NMR spectroscopy
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Clear colorless Liquid
  • Residue after evaporation:
    ≤0.05 %
  • Suitability:
    (for 1H-NMR position ref.) Passes test