
4-(4-Isopropylpiperazin-1-yl)phenylboronic acid pinacol ester 97%
Catalog # ACRO445080010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to tan Solid
- Infrared spectrum:Conforms
- Melting point:136 to 142 °C
- GC:≥96.0 %
- NMR (1H-NMR):Consistent with structure
Specification Test Results
About this item
:
:330.28 g/mol
Melting Pt:139…141 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1073354-18-3
Formula:C₁₉H₃₁BN₂O₂
MDL Number:MFCD06795656