
2-Chloro-1,3-dimethylimidazolinium chloride 90%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37091-73-9
:
Formula:C₅H₁₀Cl₂N₂
MDL Number:MFCD09039290
:169.05 g/mol
Melting Pt:112…121 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to off-white to brown Crystalline powder or solid
- Infrared spectrum:Conforms
- HPLC:≥88.0 %
- Elemental Analysis:31.26 to 39.79 % (C)
Specification Test Results
Frequently Bought Together