Order Entry
Poland
ContactUsLinkComponent
(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulphonate salt 97%
Irritant
(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulphonate salt 97%
Certificates
undefined
Irritant
(S)-(-)-3-Aminotetrahydrofuran-p-toluenesulphonate salt 97%
Specification Test Results
  • Appearance:
    White to off-white to light brown Crystals or solid
  • Infrared spectrum:
    Conforms
  • Elemental Analysis:
    48.9 to 53.0 % C
  • Specific optical rotation:
    -3° to -5° (20°C, 589 nm) (c=1, methanol)