(2S,5S)-1-[2-(1,3-Dioxolan-2-yl)phenyl]-2,5-dimethylphospholane 97%
Catalog # ACRO435171000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- NMR:≥96.0 %
- HPLC:≥96.0 %
Specification Test Results
About this item
:
:264.305 g/mol
Density:1.103 g/cm³
Flash Pt:>110 °C
Storage Temperature:Ambient
CAS Number:695816-47-8
Formula:C₁₅H₂₁O₂P
MDL Number:MFCD09265097