Order Entry
Poland
ContactUsLinkComponent
3-Chloro-4-fluorophenylmagnesium bromide 0.5 M in THF, AcroSeal®
Irritant
Corrosive
Flammable
Health
3-Chloro-4-fluorophenylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 431990500.
CAS Number:  
undefined
Irritant
Corrosive
Flammable
Health
3-Chloro-4-fluorophenylmagnesium bromide 0.5 M in THF, AcroSeal®
Catalog # 431990500.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

Specification Test Results

  • Appearance:
    Yellow to brown Solution
  • Titration Grignard:
    0.40 to 0.60 M
  • Refractive index:
    1.4250 to 1.4270 (20°C, 589 nm)
  • GC:
    ≥95.0 %

Specification Test Results

About this item

 :
 :233.747 g/mol
Boiling Pt:65 °C (760 mmHg)
Density:0.966 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
UN:2924
ADR:3,II
 :GHS07
 :GHS05
 :GHS02
 :GHS08
CAS Number:413589-34-1
Formula:C₆H₃BrClFMg
MDL Number:MFCD02260212