Order Entry
Poland
ContactUsLinkComponent
5-Fluoro-2-methoxyphenylmagnesium bromide 0.5 M in THF, AcroSeal®
 
 
 
 
5-Fluoro-2-methoxyphenylmagnesium bromide 0.5 M in THF, AcroSeal®
  431880500.
 :  
undefined
 
 
 
 
5-Fluoro-2-methoxyphenylmagnesium bromide 0.5 M in THF, AcroSeal®
  431880500.
 :  

 

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    50 mL
  • Seq:
    2370

 

 

  • Appearance:
    Clear yellow to brown Solution (May develop some turbidity or precipitate)
  • Titration Grignard:
    0.40 to 0.60 M
  • Melting point:
    67 to 70 °C
  • GC:
    ≥99.0 %
  • Enantiomeric excess:
    ≥99.0 %
  • Specific optical rotation:
    +53° to +59° (20 °C, 589 nm) (c=1, CHCl3)

 

 

 :
 :0.948 g/cm³
 :–17 °C
 :3,II
 :229.331 g/mol
 :Ambient
 :2924
 :GHS07
 :GHS05
 :GHS02
 :GHS08
 :188132-02-7
 :C₇H₆BrFMgO
 :MFCD01311481