



[3-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide 0.25 M in THF, AcroSeal®
431660500.
:
- Pack type:Glass bottle, AcroSeal®
- Pk:50 mL
- Seq:2370
- Appearance:Clear brown Solution
- Titration Grignard:0.15 to 0.35 M
- GC:≥90.0 %
:
:264.449 g/mol
:0.922 g/cm³
:–17 °C
:Ambient
:2924
:3,II
:GHS08
:GHS05
:GHS02
:GHS07
:480424-79-1
:C₁₁H₁₄BrMgN
:MFCD03840892