
2-Chloro-3-formylpyridine 98%
:36404-88-3
:
:C₆H₄ClNO
:MFCD01315308
:141.56 g/mol
:47…50 °C
:Ambient
:2811
:6.1,III
:GHS07
- Appearance:Light yellow Crystalline powder or solid
- Infrared spectrum:Conforms
- Melting point:47 to 52 °C
- GC:≥97.5 %
Frequently Bought Together