Order Entry
Poland
ContactUsLinkComponent
Trimethylboroxine 50% (w/w) 3.5 M in THF, AcroSeal®
Corrosive
Flammable
Irritant
Health
Trimethylboroxine 50% (w/w) 3.5 M in THF, AcroSeal®
Catalog # 429191000.
CAS Number:  
undefined
Corrosive
Flammable
Irritant
Health
Trimethylboroxine 50% (w/w) 3.5 M in THF, AcroSeal®
Catalog # 429191000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Colorless to yellow Solution
  • Assay:
    40.0 to 60.0 % (ex Boron)
  • Refractive index:
    1.4480 to 1.4530 (20°C, 589 nm)
  • GC:
    ≥98.0 %
  • Stabilizer:
    Abt 0.1% alpha-tocopherol (synthetic)

Specification Test Results

About this item

 :
 :125.532 g/mol
Boiling Pt:78…80 °C (760 mmHg)
Melting Pt:–38 °C
Density:0.89 g/cm³
Flash Pt:–9 °C
Storage Temperature:Ambient
UN:1993
ADR:3,II
 :GHS05
 :GHS02
 :GHS07
 :GHS08
CAS Number:823-96-1
Formula:C₃H₉B₃O₃
MDL Number:MFCD00013354