Aluminium acetate basic 90%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Aluminium Acetate Monobasic
- CAS Number:142-03-0
- Formula:C₄H₇AlO₅
- MDL Number:MFCD00008688
- Storage Temperature:Ambient
2 Options Available Below
Product Details & Documents
Synonyms: Aluminium Acetate Monobasic
- CAS Number:142-03-0
- Formula:C₄H₇AlO₅
- MDL Number:MFCD00008688
- Storage Temperature:Ambient
Documents
Specifications for Product Family
Specification Test Results
- Appearance:White Powder
- Infrared spectrum:Conforms
- Aluminium (Al):15.0 to 18.3 % (ICP)
Product Family Options
Product Information
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



