
L-Methioninamide hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:16120-92-6
:
Formula:C₅H₁₃ClN₂OS
MDL Number:MFCD00039084
:184.682 g/mol
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Titration Argentometric:97.5 to 102.5 %
- NMR (1H-NMR):Consistent with structure
- Specific optical rotation:+16° to +18° (20°C, 589 nm) (c=1,H2O)
Specification Test Results
Frequently Bought Together