Order Entry
Poland
ContactUsLinkComponent
(3S)-(-)-3-(Dimethylamino)pyrrolidine 97%
Irritant
Corrosive
Flammable
(3S)-(-)-3-(Dimethylamino)pyrrolidine 97%
Catalog # ACRO426450010
CAS Number:  
undefined
Irritant
Corrosive
Flammable
(3S)-(-)-3-(Dimethylamino)pyrrolidine 97%
Catalog # ACRO426450010
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4640 to 1.4660 (20°C, 589 nm)
  • GC:
    96.0 %
  • Specific optical rotation:
    -12° to -16° (20°C, 589 nm) (c=1% ethanol)

Specification Test Results

About this item

 :
 :114.19 g/mol
Boiling Pt:164 °C (760 mmHg)
Density:0.899 g/cm³
Flash Pt:51 °C
Storage Temperature:Ambient
UN:2733
ADR:3,III
 :GHS07
 :GHS05
 :GHS02
CAS Number:132883-44-4
Formula:C₆H₁₄N₂
MDL Number:MFCD00191348