Order Entry
Poland
ContactUsLinkComponent
Trimethylsilyl cyanide 98%, AcroSeal®
Toxic
Environment
Flammable
Trimethylsilyl cyanide 98%, AcroSeal®
Catalog # 422061000.
CAS Number:  
undefined
Toxic
Environment
Flammable
Trimethylsilyl cyanide 98%, AcroSeal®
Catalog # 422061000.
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3900 to 1.3930 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

About this item

CAS Number:7677-24-9
 :
Formula:C₄H₉NSi
MDL Number:MFCD00001765
 :99.21 g/mol
Boiling Pt:114 °C (1013 hPa)
Melting Pt:11 °C
Density:0.744 g/cm³
Flash Pt:1 °C
UN:3384
ADR:6.1,I
 :GHS06
 :GHS09
 :GHS02
Storage Temperature:Refrigerator