
3-Chloro-2,4(3H,5H)-furandione 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4971-55-5
:
Formula:C₄H₃ClO₃
MDL Number:MFCD00134273
:134.52 g/mol
Melting Pt:208 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Beige Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
Specification Test Results
Frequently Bought Together