Order Entry
Poland
ContactUsLinkComponent
Titanium(III) chloride solution 20% (w/v) in hydrochloric acid 2M
Corrosive
Titanium(III) chloride solution 20% (w/v) in hydrochloric acid 2M
Certificates
undefined
Corrosive
Titanium(III) chloride solution 20% (w/v) in hydrochloric acid 2M
Certificates

Specification Test Results

  • Appearance:
    Dark purple Solution
  • Titration Complexometric:
    18 to 23 %
  • Titration with NaOH:
    1.8 to 2.2 N HCl

Specification Test Results

Related Information

Frequently Bought Together