

1,3-Dimethylbarbituric acid 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:769-42-6
:
Formula:C₆H₈N₂O₃
MDL Number:MFCD00006675
:156.14 g/mol
Melting Pt:122…125 °C
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:White to yellow to light brown Powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:121 to 125 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together