Order Entry
Poland
ContactUsLinkComponent
 
Potassium fluoride 40% (w/w) on alumina
 
undefined
Potassium fluoride 40% (w/w) on alumina
 

 

  • Appearance:
    White Powder
  • Assay:
    35 to 45 wt % (Mass balance)
  • Water:
    3 to 10 % (K.F.)

 

 

Frequently Bought Together