Order Entry
Poland
Orders LinkContactUsLinkComponent
3,3-Dimethylbutyraldehyde

3,3-Dimethylbutyraldehyde

Irritant
Flammable
3,3-Dimethylbutyraldehyde
Certificates
3,3-Dimethylbutyraldehyde

3 Options Available Below

Product Details & Documents
Synonyms:  3,3-Dimethylbutanal
  • CAS Number:2987-16-8
  • Formula:C₆H₁₂O
  • MDL Number:MFCD00042807
  • Molecular Weight:100.16 g/mol
  • Boiling Pt:104…106 °C (760 mmHg)
  • Melting Pt:–24 °C
  • Density:0.798 g/cm³
  • Flash Pt:13 °C
  • Storage Temperature:Ambient
  • UN:1989
  • ADR:3,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3960 to 1.3980 (20°C, 589 nm)
  • GC:
    ≥94 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 360 zł
    360 zł
    1 * 1 g
       360 zł

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 1 200 zł
    1 200 zł
    1 * 5 g
       1 200 zł

  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 4 020 zł
    4 020 zł
    1 * 25 g
       4 020 zł

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.