Order Entry
Poland
ContactUsLinkComponent
Methacrolein 90% stabilised, AcroSeal®
 
 
 
 
 
Methacrolein 90% stabilised, AcroSeal®
  196501000.
 :  
undefined
 
 
 
 
 
Methacrolein 90% stabilised, AcroSeal®
  196501000.
 :  

 

  • Pack type:
    Glass bottle, AcroSeal®
  • Pk:
    100 mL
  • Seq:
    2440

 

 

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥88 %
  • Stabilizer:
    0.1 to 0.5 % Hydroquinone

 

 

 :78-85-3
 :
 :C₄H₆O
 :MFCD00006974
 :70.09 g/mol
 :69 °C (760 mmHg)
 :–81 °C
 :0.847 g/cm³
 :–15 °C
 :2396
 :3,II
 :GHS06
 :GHS05
 :GHS02
 :GHS08
 :GHS09
 :Refrigerator