Order Entry
Poland
ContactUsLinkComponent
Mercury (II) oxide, red 99+% ACS
Toxic
Health
Environment
Mercury (II) oxide, red 99+% ACS
Certificates
undefined
Toxic
Health
Environment
Mercury (II) oxide, red 99+% ACS

Specification Test Results

  • Appearance:
    Orange-red to red Powder
  • Titration Complexometric:
    ≥99.0 %
  • Chloride (Cl):
    ≤0.025 %
  • Sulfate (SO4):
    ≤0.015 %
  • Iron (Fe):
    ≤0.005 %
  • Insoluble matter:
    ≤0.03 % (in dilute HCl)
  • Residue after reduction:
    ≤0.025 %
  • Nitrogen compounds (as N):
    ≤0.005 %

Specification Test Results

Related Information

Frequently Bought Together