Order Entry
Poland
ContactUsLinkComponent
Aluminium ammonium sulphate dodecahydrate ACS
Irritant
Aluminium ammonium sulphate dodecahydrate ACS
Catalog # ACRO192382500
CAS Number:  
undefined
Irritant
Aluminium ammonium sulphate dodecahydrate ACS
Catalog # ACRO192382500
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g
  • Seq:
    3545

Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Titration Complexometric:
    98.0 to 102.0 %
  • Chloride (Cl):
    ≤0.001 %
  • Calcium (Ca):
    ≤0.05 %
  • Iron (Fe):
    ≤0.001 %
  • Potassium (K):
    ≤0.05 %
  • Sodium (Na):
    ≤0.01 %
  • Heavy metals (as Pb):
    ≤0.001 %
  • Insoluble matter:
    ≤0.005 %

Specification Test Results

About this item

 :
 :453.33 g/mol
Boiling Pt:200 °C (760 mmHg)
Melting Pt:93.5 °C
Density:1.64 g/cm³
 :GHS07
Storage Temperature:Ambient
CAS Number:7784-26-1
 :MFCD00149958
Formula:AlNH₄(SO₄)₂·12H₂O