Order Entry
Poland
ContactUsLinkComponent
Decylamine 99%
Decylamine 99%
Certificates
undefined
Decylamine 99%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4350 to 1.4370 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together