Order Entry
Poland
ContactUsLinkComponent
KP-Mega moulds
KP-Mega moulds
Catalog # KLIN3055
Supplier:  KLINIPATH
KP-Mega moulds
Catalog # KLIN3055
Supplier:  KLINIPATH
Specifications
  • Description:
    Mega moulds
  • Dimensions:
    33×24×12 mm
  • Pk:
    6
Specifications
About this item
These moulds are made of stainless steel and are suitable for Mega cassettes.