HyClone™ Ham's F-12 Basal Media, Cell Culture Media, Liquid
Supplier: HyClone products (Cytiva)
Certificates
About this item
Ham's F-12 media was originally derived by modifying Ham's F-10 to include increased concentrations of choline, inositol, putrescine, and several amino acids. It was developed specifically for serum free culturing of Chinese Hamster Ovary (CHO) cells and has also been used with supplementation to grow chondrocytes, primary rat hepatocytes, rat prostate epithelial cells, carcinoma cells, rat skeletal myoblasts and rat, rabbit, and chicken embryos.
- Hydrated using WFI quality water
- 0,1 μm sterile filtered
- Manufactured using ISO 9001:2000 certified processes
A Certificate of Analysis is included in each package with preparation instructions, intended use, and handling and performance criteria.
Ordering information: Liquid media are supplied in unbreakable, square PETG bottles.
Frequently Bought Together