Jars, wide mouth, depyrogenated glass
Supplier: EP SCIENTIFIC
Certificates
About this item
Clear or amber glass with closure.
- Packaging and storing articles that will be terminally sterilized
- Storage of laboratory reagents and medias
- Sample storage and water sampling
- Meet endotoxin levels of less than 0,06 EU/ml
Delivery: It is supplied with certificate of analysis.
Specifications
- Catalog No:
- EPSC132-16C/PF
- EPSC130-04A/PF
- EPSC130-04C/TL/PF
- EPSC130-04C/PF
- EPSC131-08C/PF
- EPSC131-08A/PF
- EPSC130-02C/PF
- EPSCC20-02A/PF
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement
- NAR Certificate of Retirement