

Copper(II) sulphate, anhydrous 98%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7758-98-7
:
Formula:CuSO₄
MDL Number:MFCD00010981
:159.61 g/mol
Melting Pt:200 °C (dec.)
Density:3,603 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:3077
ADR:9,III
:GHS07
:GHS09
Specification Test Results
- Appearance:Light grey Powder
- Titration Iodometric:≥97.5 %
- Chloride (Cl):≤50 ppm
- Iron (Fe):≤100 ppm
Specification Test Results
Frequently Bought Together