


Zirconium(IV) Propoxide ∼ 70% in propanol, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:23519-77-9
:
Formula:C₁₂H₂₈O₄Zr
MDL Number:MFCD00015307
:327.58 g/mol
Density:1.044 g/cm³
Flash Pt:23 °C
UN:1993
ADR:3,III
:GHS07
:GHS05
:GHS02
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to yellow to amber Solution
- Assay:27.0 to 29.0 % (ZrO2)
- Melting point:73 to 80 °C
- GC:≥97.5 %
Specification Test Results
Related Information
Certificates
Search by Lot NumberACRO42793
SDS
ACRO427931000_TDS_EN
Product specification
AcroSeal Brochure VWR Final AO_AcroSeal_Product Lists_VWR_final
Product Literature
Frequently Bought Together