



Isopropenylmagnesium bromide solution 0.5 M in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13291-18-4
:
Formula:C₃H₅BrMg
MDL Number:MFCD00192240
:145.282 g/mol
Boiling Pt:67 °C (760 mmHg)
Density:0.927 g/cm³
Flash Pt:2 °C
UN:3399
ADR:4.3,I
:GHS07
:GHS05
:GHS02
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow to grey to brown Solution
- Titration Active base:0.45 to 0.60 M
- Melting point:37 to 41 °C
- GC:≥96.0 %
Specification Test Results
Related Information
Frequently Bought Together