5-Bromo-1H-indole-3-carbaldehyde 97%
ACRO399790050
:
- Pack type:Glass bottle
- Pk:5 g
- Seq:3173
- Appearance:White to pink to light brown Powder
- Infrared spectrum:Conforms
- Melting point:200 to 207 °C
- HPLC:≥96.0 %
:
:205…206 °C
:224.06 g/mol
:GHS07
:Ambient
:877-03-2
:C₉H₆BrNO
:MFCD00152016