



3,5-Bis(trifluoromethyl)phenylmagnesium bromide 0.5 M in THF, AcroSeal®
392151000.
:
- Pack type:Glass bottle, AcroSeal®
- Pk:100 mL
- Seq:2440
- Appearance:Dark brown Solution
- Titration Grignard:0.40 to 0.60 M
:
:317.311 g/mol
:65 °C (760 mmHg)
:1.015 g/cm³
:–17 °C
:Ambient
:2924
:3,II
:GHS05
:GHS02
:GHS07
:GHS08
:112981-69-8
:C₈H₃BrF₆Mg
:MFCD07369723