Diphenhydramine hydrochloride 99%
:147-24-0
:
:C₁₇H₂₁NO
:MFCD00012479
:291.82 g/mol
:168…172 °C
:GHS07
:Ambient
- Appearance:White Adhering crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:168 to 172 °C
- Titration Argentometric:≥98.5 %
Frequently Bought Together