Order Entry
Poland
ContactUsLinkComponent
4-Dodecyldiethylenetriamine 80% tech.
Corrosive
4-Dodecyldiethylenetriamine 80% tech.
Catalog # ACRO388210250
CAS Number:  
undefined
Corrosive
4-Dodecyldiethylenetriamine 80% tech.
Catalog # ACRO388210250
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • GC:
    ≥75 %

Specification Test Results

About this item

 :
 :271.493 g/mol
Boiling Pt:140 °C (.2 mmHg)
Melting Pt:22 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:2735
ADR:8,III
 :GHS05
CAS Number:4182-44-9
Formula:C₁₆H₃₇N₃
MDL Number:MFCD00041897