
Piperonyloyl chloride 98%
Catalog # ACRO370790010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to yellow to light beige Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Melting point:77 to 81 °C
- GC:≥97.5 %
Specification Test Results
About this item
:
:184.575 g/mol
Melting Pt:77…81 °C
Storage Temperature:Ambient
UN:3261
ADR:8,II
:GHS05
CAS Number:25054-53-9
Formula:C₈H₅ClO₃
MDL Number:MFCD00016904