Order Entry
Poland
ContactUsLinkComponent
Triolein 99%
Triolein 99%
Certificates
undefined
Triolein 99%

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4680 to 1.4700 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together