

2-(Piperazin-1-yl)pyrazine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34803-68-4
:
Formula:C₈H₁₂N₄
MDL Number:MFCD00040766
:164.212 g/mol
Boiling Pt:105…108 °C (.5mbar)
UN:3259
ADR:8,III
:GHS07
:GHS05
Storage Temperature:Ambient
Specification Test Results
- Appearance:Yellow Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together