
N-(3-Trifluoromethylphenyl)anthranilic acid 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:530-78-9
:
Formula:C₁₄H₁₀F₃NO₂
MDL Number:MFCD00002422
:281.23 g/mol
Melting Pt:132…135 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Specification Test Results
- Appearance:White to light yellow to grey-green Crystalline powder and/or chunks
- Infrared spectrum:Conforms
- Melting point:132 to 137 °C
- Titration with NaOH:96.0 to 104.0 %
- Residue after ignition:≤0.5 %
- Specific optical rotation:-91.0° to -93.5° (20 °C, 589 nm)
- Acidity:Passes test
- Color of solution:Passes test
- Chloride (Cl):≤0.018 %
- Sulfate (SO4):≤0.025 %
- Arsenic (As):≤1 ppm
- Ca and Mg:≤0.005 % (as Ca)
- Hydroxymethylfurfural:Passes test
Specification Test Results
Frequently Bought Together