Order Entry
Poland
ContactUsLinkComponent
3,5-Pyrazoledicarboxylic acid monohydrate 97%
Irritant
3,5-Pyrazoledicarboxylic acid monohydrate 97%
Certificates
undefined
Irritant
3,5-Pyrazoledicarboxylic acid monohydrate 97%
Specification Test Results
  • Appearance:
    White to off-white Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Water:
    8.0 to 11.0 %
  • Titration with NaOH:
    96.0 to 104.0 %