
N-(Methoxymethyl)-N-(trimethylsilylmethyl)benzylamine 96%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:93102-05-7
:
Formula:C₁₃H₂₃NOSi
MDL Number:MFCD00674005
:237.42 g/mol
Boiling Pt:76 °C (.3 mmHg)
Density:0.928 g/cm³
Flash Pt:66 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4910 to 1.4940 (20°C, 589 nm)
- Elemental Analysis:≥95 %
Specification Test Results
Frequently Bought Together