6-Azabicyclo[3.2.0]heptan-7-one 98%
Catalog # ACRO335835000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mg
- Seq:2725
Specifications
Specification Test Results
- Appearance:Off-white to light yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:45 to 50 °C
- Elemental Analysis:≥97.5 %
Specification Test Results
About this item
:
Melting Pt:45…50 °C
:111.144 g/mol
Storage Temperature:Ambient
CAS Number:22031-52-3
Formula:C₆H₉NO
MDL Number:MFCD01863277