Order Entry
Poland
ContactUsLinkComponent
Methyl-2-(2-chloro-4-fluorophenyl)acetate 98%
Methyl-2-(2-chloro-4-fluorophenyl)acetate 98%
Catalog # ACRO304780050
CAS Number:  
undefined
Methyl-2-(2-chloro-4-fluorophenyl)acetate 98%
Catalog # ACRO304780050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5060 to 1.5080 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

About this item

 :
 :202.609 g/mol
Storage Temperature:Ambient
CAS Number:214262-88-1
Formula:C₉H₈ClFO₂
MDL Number:MFCD00800611