Order Entry
Poland
ContactUsLinkComponent
Hydrogen peroxide-urea 35% H₂O₂/g (w/w) stabilised
Corrosive
Oxidizing
Hydrogen peroxide-urea 35% H₂O₂/g (w/w) stabilised
Certificates
undefined
Corrosive
Oxidizing
Hydrogen peroxide-urea 35% H₂O₂/g (w/w) stabilised
Specification Test Results
  • Appearance:
    White Tablets
  • Stabilizer:
    1 % Sodium pyrophosphate
  • Titration with KMnO4:
    ≥33.5 % (H2O2)