Order Entry
Poland
ContactUsLinkComponent
Avidin, Pierce™
Avidin, Pierce™
  PIER21121
 :  
undefined
Avidin, Pierce™
  PIER21121
 :  
  • Pk:
    10 mg
  • Description:
    Avidin
Pierce™ Avidin is used in biotin-based detection methods, including Western blotting or ELISA detection with appropriate enzyme substrates. It is a purified protein from egg whites and it is stable over wide range of pH and temperature.
  • Native avidin - glycoprotein from chicken egg whites, 67 kDa, pI = 10
  • Glycosylation and isoelectric point renders avidin more soluble than streptavidin
  • Significantly less expensive than recombinant streptavidin
  • Avidin is usually preferred over streptavidin for avidin-biotin complex (ABC) staining methods in immunohistochemistry (IHC)