Order Entry
Poland
Orders LinkContactUsLinkComponent
VWR®, COD calibration standards
VWR®, COD calibration standards
Supplier:  VWR Chemicals
Certificates
VWR®, COD calibration standards
VWR®, COD calibration standards
17 Options Available Below
Product Details & Documents
These standards are ideal to use as control standards for COD measurements to verify that correct analysis for COD has taken place.
Achieving an acceptable result for a control standard will improve confidence in sample readings for this important environmental parameter.
Product Family Options
Product Information
  • Concentration
    Pack type
    Pk
    Availability
    Price
  • 85050.260
    COD standard solution
    COD standard solution
    10 mg/L
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85053.260
    COD standard solution
    COD standard solution
    20 mg/L
    Glass bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85063.260
    COD standard solution
    COD standard solution
    50 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85059.260
    COD standard solution
    COD standard solution
    100 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85062.260
    COD standard solution
    COD standard solution
    200 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85064.260
    COD standard solution
    COD standard solution
    500 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85066.260
    COD standard solution
    COD standard solution
    600 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85060.260
    COD standard solution
    COD standard solution
    1000 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85051.260
    COD standard solution
    COD standard solution
    1300 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85052.260
    COD standard solution
    COD standard solution
    1500 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85054.260
    COD standard solution
    COD standard solution
    2000 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85065.260
    COD standard solution
    COD standard solution
    5000 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85057.260
    COD standard solution
    COD standard solution
    6000 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85061.260
    COD standard solution
    COD standard solution
    10000 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85055.260
    COD standard solution
    COD standard solution
    20000 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85056.260
    COD standard solution
    COD standard solution
    30000 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł
  • 85058.260
    COD standard solution
    COD standard solution
    60000 mg/L
    Plastic bottle
    500 mL
    Log in for availability
    Log in for availability
    1 * 500 mL
    1 * 500 mL - 727 zł
    727 zł
    1 * 500 mL
       727 zł