Calf alkaline phosphatase (from intestine), Pierce™
Supplier: Thermo Fisher Scientific
Certificates
About this item
This purified calf intestinal alkaline phosphatase (CIP) is supplied in Tris buffer and 50% glycerol for use in protein research methods. The main applications for alkaline phosphatase in molecular biology and protein research are to remove 5'-phosphate groups from DNA or as a reporter system for immunoassays such as ELISA. For these latter methods, the enzyme is usually conjugated to specific primary or secondary antibodies and its activity is detected with a color-forming (or light-generating) substrate.
- Purified form - ready to dilute and conjugate without prior dialysis
- Concentrated - approximately 20 mg/ml (lot-specific value reported)
- High specific activity - typically greater than 1600 units/mg (lot-specific value reported)
- Tris buffered solution - supplied in 5 mM Tris, 5 mM magnesium chloride and 0.1 mM zinc chloride, pH ~7.0 in 50% glycerol
One unit equals the amount of protein required to hydrolyze one micromole of p-nitrophenyl phosphate (PNPP) per minute at 25 °C in a glycine buffer, pH 9.6.
Caution: Store at 2–8 °C. For Research Use Only. Not for use in diagnostic procedures.
Specifications
- Source:Intestine
- Species:Calf
- Storage conditions:2–8 °C
- Enzyme name:Alkaline phosphatase
- Enzyme synonyms:CIP
- Solubility:1600 units/mg
- Concentration:20 mg/ml
Specifications
Frequently Bought Together