Order Entry
Poland
ContactUsLinkComponent
 
Alizarin complexone dihydrate for fluoride determination, Supelco®
  1.01010.0005
 :  Merck
 :  
Alizarin complexone dihydrate for fluoride determination, Supelco®
  1.01010.0005
 :  Merck
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

 

 

  • Solubility:
    (20 °C) practically insoluble
  • Molar Mass:
    421.36 g/mol
  • Bulk Density:
    540 kg/m3

 

 

 :
 :421.36 g/mol
 :180 °C
 :M-Clarity™ quality level = MQ100
 :Ambient
 :223-544-2
 :455303-00-1
 :C₁₉H₁₅NO₈·2H₂O
 :MFCD00149067