5(6)-Carboxytetramethylrhodamine succinimidyl ester [5(6)-TAMRA SE] mixture of isomers fluorescent dye, Pierce™
Catalog # PIER46406
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:527.52 g/mol
Storage Temperature:Freezer
CAS Number:246256-50-8
Formula:C₂₉H₂₅N₃O₇
MDL Number:MFCD00077324