Potassium citrate monohydrate 99.0-100.5%, granular USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:6100-05-6
:
Formula:HOC(COOK)(CH₂COOK)₂·H₂O
MDL Number:MFCD00150442
Merck Index:13,07706
Melting Pt:230 °C
Density:1.98 g/cm³ (20 °C)
:324.41 g/mol
EINECS:212-755-5
Specification Test Results
- Assay:99% Min.
Specification Test Results
Frequently Bought Together