

1H,1H-Heptafluoro-n-butyl methacrylate 97% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13695-31-3
:
Formula:C₈H₇F₇O₂
MDL Number:MFCD00039251
:268.13 g/mol
Boiling Pt:134…136 °C
Density:1,345
Flash Pt:36 °C (96 °F)
EINECS:237-217-7
ADR:3,III
:GHS02
:GHS07
Related Information
Frequently Bought Together