Order Entry
Poland
Orders LinkContactUsLinkComponent
D-Amygdalin hydrate 96%

D-Amygdalin hydrate 96%

D-Amygdalin hydrate 96%
Certificates
D-Amygdalin hydrate 96%

2 Options Available Below

Product Details & Documents

Purity: 96% CAS No.: 29883-15-6 Molecular Formula: C20H27NO11.xH2O Formula Weight: 457.44(anhy) Melting Point: 215-220° Beilstein Reference: 66856 Optical Rotation: -36° (c=1.2 in water) MDL No.: MFCD00006598

Synonyms:  D-Mandelonitrile-beta-D-glucosido-6-beta-D-glucoside hydrate, Amygdalin hydrate
  • CAS Number:29883-15-6
  • Formula:C₂₀H₂₇NO₁₁·xH₂O
  • MDL Number:MFCD00006598
  • Molecular Weight:457.44 g/mol (anhy)
  • Merck Index:14,597
  • Melting Pt:215…220 °C
  • Storage Temperature:Ambient
  • EINECS:249-925-3
Specifications for Product Family

Specification Test Results

  • Appearance (Color):
    White
  • Form:
    Crystalline solid
  • Assay (HPLC):
    >95.0%
  • Loss on Drying:
    <10.0% @ 110°C
  • Optical Rotation:
    -38 ±2° (c=1.2 in water) on dry wt. basis
Product Family Options

Product Information